Thermo Scientific Chemicals

2-Chloro-5-methoxyaniline, 98%, Thermo Scientific Chemicals

Catalog number: 453780050
5 g, Each
Thermo Scientific Chemicals

2-Chloro-5-methoxyaniline, 98%, Thermo Scientific Chemicals

Catalog number: 453780050
5 g, Each
Quantity
Catalog number: 453780050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2401-24-3
IUPAC Name
2-chloro-5-methoxyaniline
Molecular Formula
C7H8ClNO
InChI Key
GBOUQGUQUUPGLO-UHFFFAOYSA-N
SMILES
COC1=CC=C(Cl)C(N)=C1
Appearance (Color)
Colorless to yellow to brown
Appearance (Form)
Low melting solid or liquid
Infrared spectrum
Conforms
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text