Thermo Scientific Chemicals

Cadmium bromide, 98%, pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 453860250
25 g, Each
Thermo Scientific Chemicals

Cadmium bromide, 98%, pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 453860250
25 g, Each
Quantity
Catalog number: 453860250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7789-42-6
Appearance (Color)
White to light yellow
Appearance (Form)
Crystalline powder
Assay
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text