Cadmium bromide tetrahydrate, 98%, pure, Thermo Scientific Chemicals
Cadmium bromide tetrahydrate, 98%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cadmium bromide tetrahydrate, 98%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 453870250
Price (USD)/ Each
27.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
32.60 
Save 4.95 (15%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
27.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
32.60 
Save 4.95 (15%)
Add to cart
Cadmium bromide tetrahydrate, 98%, pure, Thermo Scientific Chemicals
Catalog number453870250
Price (USD)/ Each
27.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
32.60 
Save 4.95 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namecadmium(2+) tetrahydrate dibromide
Molecular FormulaBr2CdH8O4
InChI KeyPANYFCXNZSEXSU-UHFFFAOYSA-L
SMILESO.O.O.O.[Br-].[Br-].[Cd++]
Molecular Weight (g/mol)344.28
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder or crystals
Assay>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only