Thermo Scientific Chemicals

Cadmium bromide tetrahydrate, 98%, pure, Thermo Scientific Chemicals

Catalog number: 453871000
100 g, Each
Thermo Scientific Chemicals

Cadmium bromide tetrahydrate, 98%, pure, Thermo Scientific Chemicals

Catalog number: 453871000
100 g, Each
Quantity
Catalog number: 453871000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13464-92-1
IUPAC Name
cadmium(2+) tetrahydrate dibromide
Molecular Formula
Br2CdH8O4
InChI Key
PANYFCXNZSEXSU-UHFFFAOYSA-L
SMILES
O.O.O.O.[Br-].[Br-].[Cd++]
Appearance (Color)
White
Appearance (Form)
Powder or crystals
Assay
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text