Thermo Scientific Chemicals

Cobalt(II) hydroxide, 95%, pure, Thermo Scientific Chemicals

Catalog number: 453892500
250 g, Each
Thermo Scientific Chemicals

Cobalt(II) hydroxide, 95%, pure, Thermo Scientific Chemicals

Catalog number: 453892500
250 g, Each
Quantity
Catalog number: 453892500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
21041-93-0
IUPAC Name
λ²-cobalt(2+) dihydroxide
Molecular Formula
CoH2O2
InChI Key
ASKVAEGIVYSGNY-UHFFFAOYSA-L
SMILES
[OH-].[OH-].[Co++]
Appearance (Form)
Powder
Titration excess HCl
>=94 %
Appearance (Color)
Pink to red

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text