L-Phenylalanine, 98.5-101.0%, specified according to the requirements of USP
L-Phenylalanine, 98.5-101.0%, specified according to the requirements of USP
L-Phenylalanine, 98.5-101.0%, specified according to the requirements of USP
L-Phenylalanine, 98.5-101.0%, specified according to the requirements of USP
Thermo Scientific Chemicals

L-Phenylalanine, 98.5-101.0%, specified according to the requirements of USP

CAS: 63-91-2 | C9H11NO2 | 165.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 453931000
Price (USD)
84.60
Each
In stock
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
84.60
Each
Add to cart
Chemical Identifiers
CAS63-91-2
IUPAC Name2-amino-3-phenylpropanoic acid
Molecular FormulaC9H11NO2
InChI KeyCOLNVLDHVKWLRT-UHFFFAOYNA-N
SMILESNC(CC1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Titration with HClO498.5 to 101.5 % (On dried base)
Sulfated ash=<0.4 %
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0464225Certificate of AnalysisJul 23, 2024453930250, 453931000
A0453356Certificate of AnalysisAug 25, 2023453930250, 453931000
A0341009Certificate of AnalysisMay 09, 2023453930250, 453931000
E04X026Certificate of AnalysisNov 22, 2021453930250, 453931000
A0435979Certificate of AnalysisNov 10, 2021453930250, 453931000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection