Copper(II) bromide, 99%, for analysis, anhydrous
Copper(II) bromide, 99%, for analysis, anhydrous
Copper(II) bromide, 99%, for analysis, anhydrous
Copper(II) bromide, 99%, for analysis, anhydrous
Thermo Scientific Chemicals

Copper(II) bromide, 99%, for analysis, anhydrous

CAS: 7789-45-9 | Br2Cu | 223.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 453952500
Price (EUR)
52,87
Online Exclusive
62,20
Save 9,33 (15%)
Each
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (EUR)
52,87
Online Exclusive
62,20
Save 9,33 (15%)
Each
Add to cart
Chemical Identifiers
CAS7789-45-9
IUPAC Namecopper(2+) dibromide
Molecular FormulaBr2Cu
InChI KeyQTMDXZNDVAMKGV-UHFFFAOYSA-L
SMILES[Cu++].[Br-].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver grey to black
Appearance (Form)Crystalline powder
Titration Iodometric>=98.5 %
Loss on drying=<1 %
Iron (Fe)=<0.01 %
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Acute oral toxicity
Category 4
Skin Corrosion/Irritation
Category 1
Serious Eye Damage/Eye Irritation
Category 1
Acute aquatic toxicity
Category 1
Chronic aquatic toxicity
Category 1
Label Elements
Signal Word

Danger

Hazard Statements

H302 - Harmful if swallowed

H314 - Causes severe skin burns and eye damage

H410 - Very toxic to aquatic life with long lasting effects

Precautionary Statements

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P301 + P330 + P331 - IF SWALLOWED: Rinse mouth. Do NOT induce vomiting

P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

P310 - Immediately call a POISON CENTER or doctor/physician

Share catalog number, name or link

1x1 image pixel for data collection