Thermo Scientific Chemicals

Copper(II) nitrate trihydrate, 99%, pure, Thermo Scientific Chemicals

Catalog number: 453960050
5 kg, Each
Thermo Scientific Chemicals

Copper(II) nitrate trihydrate, 99%, pure, Thermo Scientific Chemicals

Catalog number: 453960050
5 kg, Each
Quantity
Catalog number: 453960050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10031-43-3
IUPAC Name
copper(2+) dinitrate
Molecular Formula
CuN2O6
InChI Key
XTVVROIMIGLXTD-UHFFFAOYSA-N
SMILES
[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Appearance (Color)
Dark blue
Appearance (Form)
Crystalline powder or crystals
Titration Iodometric
98.5 to 104 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text