Lithium bromide, 99%, pure, anhydrous
Lithium bromide, 99%, pure, anhydrous
Lithium bromide, 99%, pure, anhydrous
Lithium bromide, 99%, pure, anhydrous
Lithium bromide, 99%, pure, anhydrous
Thermo Scientific Chemicals

Lithium bromide, 99%, pure, anhydrous

CAS: 7550-35-8 | BrLi | 86.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
2.5 kg
Packaging:
Plastic Bottle
Catalog number 453982500
Price (USD)
79.65
Special offer
Online exclusive
Ends: 26-Sep-2025
98.80
Save 19.15 (19%)
Each
-
Add to cart
Quantity:
250 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
79.65
Special offer
Online exclusive
Ends: 26-Sep-2025
98.80
Save 19.15 (19%)
Each
Add to cart
Chemical Identifiers
CAS7550-35-8
IUPAC Namelithium(1+) bromide
Molecular FormulaBrLi
InChI KeyAMXOYNBUYSYVKV-UHFFFAOYSA-M
SMILES[Li+].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Titration Argentometric>=98.5 %
pH6 to 12 (4 % in water)

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link