Bismuth(III) sulfide, 99%, pure, Thermo Scientific Chemicals
Bismuth(III) sulfide, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bismuth(III) sulfide, 99%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 453991000
Price (USD)/ Each
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS1345-07-9
IUPAC Namedibismuth(3+) trisulfanediide
Molecular FormulaBi2S3
InChI KeyYNRGZHRFBQOYPP-UHFFFAOYSA-N
SMILES[S--].[S--].[S--].[Bi+3].[Bi+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Complexometric80.1 to 82.5 % (Bi)
Appearance (Form)Powder and/or chunks
Appearance (Color)Grey

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only