Bismuth(III) nitrate pentahydrate, 98%, pure, Thermo Scientific Chemicals
Bismuth(III) nitrate pentahydrate, 98%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bismuth(III) nitrate pentahydrate, 98%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
1 kg
Catalog number 454110050
Price (USD)/ Each
416.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
490.00 
Save 73.35 (15%)
-
Add to cart
Quantity:
5 kg
Request bulk or custom format
Price (USD)/ Each
416.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
490.00 
Save 73.35 (15%)
Add to cart
Bismuth(III) nitrate pentahydrate, 98%, pure, Thermo Scientific Chemicals
Catalog number454110050
Price (USD)/ Each
416.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
490.00 
Save 73.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namebismuth(3+) pentahydrate trinitrate
Molecular FormulaBiH10N3O14
InChI KeyFBXVOTBTGXARNA-UHFFFAOYSA-N
SMILESO.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Molecular Weight (g/mol)485.07
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystals
Titration Complexometric>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only