2-Methoxypyridine-4-boronic acid, 95%, Thermo Scientific Chemicals
2-Methoxypyridine-4-boronic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methoxypyridine-4-boronic acid, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number 454370010
Price (USD)/ Each
514.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
514.00
Add to cart
2-Methoxypyridine-4-boronic acid, 95%, Thermo Scientific Chemicals
Catalog number454370010
Price (USD)/ Each
514.00
-
Add to cart
Chemical Identifiers
CAS762262-09-9
IUPAC Name(2-methoxypyridin-4-yl)boronic acid
Molecular FormulaC6H8BNO3
InChI KeyDHQMUJSACXTPEA-UHFFFAOYSA-N
SMILESCOC1=NC=CC(=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
HPLC>=94.0 %
Infrared spectrumConforms
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only