Thermo Scientific Chemicals

2-Methoxypyridine-4-boronic acid, 95%, Thermo Scientific Chemicals

Catalog number: 454372500
250 mg, Each
Thermo Scientific Chemicals

2-Methoxypyridine-4-boronic acid, 95%, Thermo Scientific Chemicals

Catalog number: 454372500
250 mg, Each
Quantity
Catalog number: 454372500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
762262-09-9
IUPAC Name
(2-methoxypyridin-4-yl)boronic acid
Molecular Formula
C6H8BNO3
InChI Key
DHQMUJSACXTPEA-UHFFFAOYSA-N
SMILES
COC1=NC=CC(=C1)B(O)O
Appearance (Color)
White to light yellow
HPLC
>=94.0 %
Infrared spectrum
Conforms
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text