6-Methylpyridine-3-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals
6-Methylpyridine-3-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Methylpyridine-3-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
Catalog number 454390010
Price (USD)/ Each
707.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
707.00
Add to cart
6-Methylpyridine-3-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals
Catalog number454390010
Price (USD)/ Each
707.00
-
Add to cart
Chemical Identifiers
CAS610768-32-6
IUPAC Name2-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular FormulaC12H18BNO2
InChI KeyJEECGGXGHJUYMN-UHFFFAOYSA-N
SMILESCC1=NC=C(C=C1)B1OC(C)(C)C(C)(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Waxy solid
NMR (1H-NMR)Consistent with structure
Appearance (Color)White to off-white
Infrared spectrumConforms
GC>=94.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only