5-Thiazolecarboxylic acid, 98%, Thermo Scientific Chemicals
5-Thiazolecarboxylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Thiazolecarboxylic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 454540010
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS14527-41-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)White to yellow to light brown
Infrared spectrumConforms
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only