Thermo Scientific Chemicals

Decamethylcyclopentasiloxane, 97%, Thermo Scientific Chemicals

Catalog number: 454580500
50 g, Each
Thermo Scientific Chemicals

Decamethylcyclopentasiloxane, 97%, Thermo Scientific Chemicals

Catalog number: 454580500
50 g, Each
Quantity
Catalog number: 454580500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
541-02-6
IUPAC Name
decamethyl-1,3,5,7,9,2,4,6,8,10-pentaoxapentasilecane
Molecular Formula
C10H30O5Si5
InChI Key
XMSXQFUHVRWGNA-UHFFFAOYSA-N
SMILES
C[Si]1(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O1
Refractive index
1.3970 to 1.3990 (20°C, 589 nm)
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text