2-Bromo-1H-imidazole, 98%, Thermo Scientific Chemicals
2-Bromo-1H-imidazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromo-1H-imidazole, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 454630050
Price (USD)/ Each
792.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
792.00
Add to cart
2-Bromo-1H-imidazole, 98%, Thermo Scientific Chemicals
Catalog number454630050
Price (USD)/ Each
792.00
-
Add to cart
Chemical Identifiers
CAS16681-56-4
IUPAC Name2-bromo-1H-imidazole
Molecular FormulaC3H3BrN2
InChI KeyAXHRGVJWDJDYPO-UHFFFAOYSA-N
SMILESBrC1=NC=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light brown
Infrared spectrumConforms
HPLC>=97.5 %
NMR (1H-NMR)Consistent with structure
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only