4-(Hydroxymethyl)benzonitrile, 97%, Thermo Scientific Chemicals
4-(Hydroxymethyl)benzonitrile, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-(Hydroxymethyl)benzonitrile, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 454650010
Price (USD)/ Each
67.10
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
67.10
Add to cart
4-(Hydroxymethyl)benzonitrile, 97%, Thermo Scientific Chemicals
Catalog number454650010
Price (USD)/ Each
67.10
-
Add to cart
Chemical Identifiers
CAS874-89-5
IUPAC Name4-(hydroxymethyl)benzonitrile
Molecular FormulaC8H7NO
InChI KeyXAASLEJRGFPHEV-UHFFFAOYSA-N
SMILESOCC1=CC=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point38°C to 44°C
Infrared spectrumConforms
GC>=96.0 %
Appearance (Color)Colorless to white to yellow
Appearance (Form)Low melting solid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only