4-Methylbenzylsulfonyl chloride, 97%, Thermo Scientific Chemicals
4-Methylbenzylsulfonyl chloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methylbenzylsulfonyl chloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 454840050
Price (USD)/ Each
210.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
210.00
Add to cart
4-Methylbenzylsulfonyl chloride, 97%, Thermo Scientific Chemicals
Catalog number454840050
Price (USD)/ Each
210.00
-
Add to cart
Chemical Identifiers
CAS51419-59-1
IUPAC Name(4-methylphenyl)methanesulfonyl chloride
Molecular FormulaC8H9ClO2S
InChI KeyJALKUHLLMWYIAT-UHFFFAOYSA-N
SMILESCC1=CC=C(CS(Cl)(=O)=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or solid
HPLC>=96.0 %
NMR (1H-NMR)Consistent with structure
Infrared spectrumConforms
Appearance (Color)White to tan
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only