2,2,2-Trifluoroethyl trifluoromethanesulfonate, 97%, Thermo Scientific Chemicals
2,2,2-Trifluoroethyl trifluoromethanesulfonate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2,2-Trifluoroethyl trifluoromethanesulfonate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 454970050
Price (USD)/ Each
152.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
152.00
Add to cart
2,2,2-Trifluoroethyl trifluoromethanesulfonate, 97%, Thermo Scientific Chemicals
Catalog number454970050
Price (USD)/ Each
152.00
-
Add to cart
Chemical Identifiers
CAS6226-25-1
IUPAC Name2,2,2-trifluoroethyl trifluoromethanesulfonate
Molecular FormulaC3H2F6O3S
InChI KeyRTMMSCJWQYWMNK-UHFFFAOYSA-N
SMILESFC(F)(F)COS(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Form)Liquid
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only