Thermo Scientific Chemicals

2,2,2-Trifluoroethyl trifluoromethanesulfonate, 97%, Thermo Scientific Chemicals

Catalog number: 454970250
25 g, Each
Thermo Scientific Chemicals

2,2,2-Trifluoroethyl trifluoromethanesulfonate, 97%, Thermo Scientific Chemicals

Catalog number: 454970250
25 g, Each
Quantity
Catalog number: 454970250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6226-25-1
IUPAC Name
2,2,2-trifluoroethyl trifluoromethanesulfonate
Molecular Formula
C3H2F6O3S
InChI Key
RTMMSCJWQYWMNK-UHFFFAOYSA-N
SMILES
FC(F)(F)COS(=O)(=O)C(F)(F)F
GC
>=96.0 %
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless to yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text