Thermo Scientific Chemicals

3'-Azido-3'-deoxythymidine, 98%, Thermo Scientific Chemicals

Catalog number: 454980050
5 g, Each
Thermo Scientific Chemicals

3'-Azido-3'-deoxythymidine, 98%, Thermo Scientific Chemicals

Catalog number: 454980050
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 454980050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
30516-87-1
IUPAC Name
1-[(2R,4S,5S)-4-azido-5-(hydroxymethyl)oxolan-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular Formula
C10H13N5O4
InChI Key
HBOMLICNUCNMMY-CFQLRCIDNA-N
SMILES
CC1=CN([C@H]2C[C@H](N=[N+]=[N-])[C@@H](CO)O2)C(=O)NC1=O
Melting point
122°C to 126°C
Infrared spectrum
Conforms
Appearance (Form)
Powder
HPLC
>=97.5 %
Appearance (Color)
White to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text