Chemical Identifiers
CAS30516-87-1
IUPAC Name1-[(2R,4S,5S)-4-azido-5-(hydroxymethyl)oxolan-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC10H13N5O4
InChI KeyHBOMLICNUCNMMY-CFQLRCIDNA-N
SMILESCC1=CN([C@H]2C[C@H](N=[N+]=[N-])[C@@H](CO)O2)C(=O)NC1=O
View more