Thermo Scientific Chemicals

4-Nitrophenyl phosphate, 97%, Thermo Scientific Chemicals

Catalog number: 455010100
10 g, Each
Thermo Scientific Chemicals

4-Nitrophenyl phosphate, 97%, Thermo Scientific Chemicals

Catalog number: 455010100
10 g, Each
Quantity
Catalog number: 455010100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
330-13-2
IUPAC Name
(4-nitrophenoxy)phosphonic acid
Molecular Formula
C6H6NO6P
InChI Key
XZKIHKMTEMTJQX-UHFFFAOYSA-N
SMILES
OP(O)(=O)OC1=CC=C(C=C1)[N+]([O-])=O
Appearance (Form)
Crystalline powder
HPLC
>=96.0 %
Appearance (Color)
White to light yellow
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text