Thermo Scientific Chemicals

Amoxicillin, 96%

Catalog number: 455140250
25 g, Each
Thermo Scientific Chemicals

Amoxicillin, 96%

Catalog number: 455140250
25 g, Each
Quantity
Catalog number: 455140250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
26787-78-0
IUPAC Name
(2S,5R,6R)-6-[(2R)-2-amino-2-(4-hydroxyphenyl)acetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate
Molecular Formula
C16H25N3O8S
InChI Key
MQXQVCLAUDMCEF-CWLIKTDRSA-N
SMILES
O.O.O.CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](N)C3=CC=C(O)C=C3)C(=O)N2[C@H]1C(O)=O
Appearance (Form)
Crystalline powder
Appearance (Color)
White to off-white
HPLC
>=95.0 %
Infrared spectrum
Conforms
Water
10 to 15 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text