Chemical Identifiers
CAS68373-14-8
IUPAC Name(2S,5R)-3,3-dimethyl-4,4,7-trioxo-4λ⁶-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Molecular FormulaC8H11NO5S
InChI KeyFKENQMMABCRJMK-RITPCOANSA-N
SMILESCC1(C)[C@@H](N2[C@@H](CC2=O)S1(=O)=O)C(O)=O
View more