Fluconazole, 98%
Thermo Scientific Chemicals

Fluconazole, 98%

 
Have Questions?
Quantity:
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 455480250
Price (USD)/ Each
896.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
1055.00 
Save 158.35 (15%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
896.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
1055.00 
Save 158.35 (15%)
Add to cart
Fluconazole, 98%
Catalog number455480250
Price (USD)/ Each
896.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
1055.00 
Save 158.35 (15%)
-
Add to cart
Chemical Identifiers
CAS86386-73-4
IUPAC Name2-(2,4-difluorophenyl)-1,3-bis(1H-1,2,4-triazol-1-yl)propan-2-ol
Molecular FormulaC13H12F2N6O
InChI KeyRFHAOTPXVQNOHP-UHFFFAOYSA-N
SMILESOC(CN1C=NC=N1)(CN1C=NC=N1)C1=CC=C(F)C=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point138°C to 142°C
Appearance (Form)Crystalline powder
HPLC>=97.5 %
Appearance (Color)White to off-white
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only