Thermo Scientific Chemicals

Arbutin, 98%, Thermo Scientific Chemicals

Catalog number: 455640050
5 g, Each
Thermo Scientific Chemicals

Arbutin, 98%, Thermo Scientific Chemicals

Catalog number: 455640050
5 g, Each
Quantity
Catalog number: 455640050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
497-76-7
IUPAC Name
2-(hydroxymethyl)-6-(4-hydroxyphenoxy)oxane-3,4,5-triol
Molecular Formula
C12H16O7
InChI Key
BJRNKVDFDLYUGJ-UHFFFAOYNA-N
SMILES
OCC1OC(OC2=CC=C(O)C=C2)C(O)C(O)C1O
Appearance (Color)
White
Infrared spectrum
Conforms
Loss on drying
=<2 %
Melting point
196°C to 201°C
Specific optical rotation
-63° to -67° (20°C, 589 nm) (c=3, H2O)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text