Thermo Scientific Chemicals

2-(Methylsulfonyl)phenol, 98%, Thermo Scientific Chemicals

Catalog number: 455750010
1 g, Each
Thermo Scientific Chemicals

2-(Methylsulfonyl)phenol, 98%, Thermo Scientific Chemicals

Catalog number: 455750010
1 g, Each
Quantity
Catalog number: 455750010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
27489-33-4
IUPAC Name
2-methanesulfonylphenol
Molecular Formula
C7H8O3S
InChI Key
ZWTZHHHSRDUVRT-UHFFFAOYSA-N
SMILES
CS(=O)(=O)C1=CC=CC=C1O
Appearance (Form)
Crystalline powder
Melting point
87°C to 92°C
Appearance (Color)
White to light yellow
Infrared spectrum
Conforms
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text