Pyrimidine-2-carboxylic acid, 95%, Thermo Scientific Chemicals
Pyrimidine-2-carboxylic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pyrimidine-2-carboxylic acid, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 455830010
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS31519-62-7
IUPAC Namepyrimidine-2-carboxylic acid
Molecular FormulaC5H4N2O2
InChI KeyZFCHNZDUMIOWFV-UHFFFAOYSA-N
SMILESOC(=O)C1=NC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to off-white
HPLC>=94.0 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only