(1-Methyl-1H-pyrazol-5-yl)methylamine, 97%, Thermo Scientific Chemicals
(1-Methyl-1H-pyrazol-5-yl)methylamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(1-Methyl-1H-pyrazol-5-yl)methylamine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 456030010
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS863548-52-1
IUPAC Name1-(1-methyl-1H-pyrazol-5-yl)methanamine
Molecular FormulaC5H9N3
InChI KeyXNTFQMKXUFFUQO-UHFFFAOYSA-N
SMILESCN1N=CC=C1CN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to brown
Appearance (Form)Liquid
Infrared spectrumConforms
Water=<1.5 %
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only