Thermo Scientific Chemicals

1,4-Cineole, 85%, Thermo Scientific Chemicals

Catalog number: 456060250
25 g, Each
Thermo Scientific Chemicals

1,4-Cineole, 85%, Thermo Scientific Chemicals

Catalog number: 456060250
25 g, Each
Quantity
Catalog number: 456060250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
470-67-7
IUPAC Name
1-methyl-4-(propan-2-yl)-7-oxabicyclo[2.2.1]heptane
Molecular Formula
C10H18O
InChI Key
RFFOTVCVTJUTAD-UHFFFAOYSA-N
SMILES
CC(C)C12CCC(C)(CC1)O2
Appearance (Color)
Clear colorless
Refractive index
1.4450 to 1.4560 (20°C, 589 nm)
Infrared spectrum
Conforms
GC
>=80 %
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text