1,10-Phenanthroline-5,6-dione, 98%
1,10-Phenanthroline-5,6-dione, 98%
1,10-Phenanthroline-5,6-dione, 98%
1,10-Phenanthroline-5,6-dione, 98%
Thermo Scientific Chemicals

1,10-Phenanthroline-5,6-dione, 98%

CAS: 27318-90-7 | C12H6N2O2 | 210.19 g/mol
Have Questions?
Catalog NumberQuantity
4560800101 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 456080010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS27318-90-7
SpecificationsSpecification SheetSpecification Sheet
HPLC>=96.0 %
Infrared spectrumConforms
Appearance (Color)Yellow to orange
Appearance (Form)Crystalline powder
Melting point255°C to 260°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection