Thermo Scientific Chemicals

Mirtazapine, 98%, Thermo Scientific Chemicals

Catalog number: 456170010
1 g, Each
Thermo Scientific Chemicals

Mirtazapine, 98%, Thermo Scientific Chemicals

Catalog number: 456170010
1 g, Each
Quantity
Catalog number: 456170010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
85650-52-8
IUPAC Name
5-methyl-2,5,19-triazatetracyclo[13.4.0.0²,⁷.0⁸,¹³]nonadeca-1(19),8,10,12,15,17-hexaene
Molecular Formula
C17H19N3
InChI Key
RONZAEMNMFQXRA-UHFFFAOYNA-N
SMILES
CN1CCN2C(C1)C1=CC=CC=C1CC1=CC=CN=C21
Appearance (Color)
White to off-white
Infrared spectrum
Conforms
HPLC
>=97.5 %
Appearance (Form)
Crystalline powder
Melting point
119°C to 123°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text