Mirtazapine, 98%
Mirtazapine, 98%
Mirtazapine, 98%
Thermo Scientific Chemicals

Mirtazapine, 98%

CAS: 85650-52-8 | C17H19N3 | 265.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 456170050
Price (USD)
663.65
Online Exclusive
737.00
Save 73.35 (10%)
Each
In stock
Add to cart
Quantity:
5 g
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
663.65
Online Exclusive
737.00
Save 73.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS85650-52-8
IUPAC Name5-methyl-2,5,19-triazatetracyclo[13.4.0.0²,⁷.0⁸,¹³]nonadeca-1(19),8,10,12,15,17-hexaene
Molecular FormulaC17H19N3
InChI KeyRONZAEMNMFQXRA-UHFFFAOYNA-N
SMILESCN1CCN2C(C1)C1=CC=CC=C1CC1=CC=CN=C21
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point119°C to 123°C
Water=<3.5 %
Infrared spectrumConforms
Appearance (Color)White to off-white
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0470723Certificate of AnalysisMay 17, 2025456170050, 456170010
A0467636Certificate of AnalysisJan 25, 2025456170050, 456170010
A0464736Certificate of AnalysisJul 24, 2024456170050, 456170010
A0406229Certificate of AnalysisMar 08, 2024456170050, 456170010
A0350375Certificate of AnalysisMar 04, 2022456170050, 456170010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection