Chemical Identifiers
IUPAC Name[(1R,2S,6S,9R)-4,4,11,11-tetramethyl-3,5,7,10,12-pentaoxatricyclo[7.3.0.0²,⁶]dodecan-6-yl]methyl sulfamate
Molecular FormulaC12H21NO8S
InChI KeyKJADKKWYZYXHBB-XBWDGYHZSA-N
SMILESCC1(C)O[C@@H]2CO[C@@]3(COS(N)(=O)=O)OC(C)(C)O[C@H]3[C@@H]2O1
Molecular Weight (g/mol)339.36
View more