Chemical Identifiers
CAS147098-20-2
IUPAC Namecalcium bis((3R,5S,6E)-7-[4-(4-fluorophenyl)-2-(N-methylmethanesulfonamido)-6-(propan-2-yl)pyrimidin-5-yl]-3,5-dihydroxyhept-6-enoate)
Molecular FormulaC44H54CaF2N6O12S2
InChI KeyLALFOYNTGMUKGG-BGRFNVSISA-L
SMILES[Ca++].CC(C)C1=NC(=NC(C2=CC=C(F)C=C2)=C1\C=C\[C@@H](O)C[C@@H](O)CC([O-])=O)N(C)S(C)(=O)=O.CC(C)C1=NC(=NC(C2=CC=C(F)C=C2)=C1\C=C\[C@@H](O)C[C@@H](O)CC([O-])=O)N(C)S(C)(=O)=O
View more