Thermo Scientific Chemicals

9-Hydroxyrisperidone, 98%

Catalog number: 456250010
1 g, Each
Thermo Scientific Chemicals

9-Hydroxyrisperidone, 98%

Catalog number: 456250010
1 g, Each
Quantity
Catalog number: 456250010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
144598-75-4
IUPAC Name
3-{2-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]ethyl}-9-hydroxy-2-methyl-4H,6H,7H,8H,9H-pyrido[1,2-a]pyrimidin-4-one
Molecular Formula
C23H27FN4O3
InChI Key
PMXMIIMHBWHSKN-UHFFFAOYNA-N
SMILES
CC1=C(CCN2CCC(CC2)C2=NOC3=CC(F)=CC=C23)C(=O)N2CCCC(O)C2=N1
Appearance (Color)
Off-white
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
168°C to 175°C
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text