D-Psicose, 98%, Thermo Scientific Chemicals
D-Psicose, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

D-Psicose, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 456270010
Price (USD)/ Each
428.65
Online exclusive
476.00 
Save 47.35 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
428.65
Online exclusive
476.00 
Save 47.35 (10%)
Add to cart
D-Psicose, 98%, Thermo Scientific Chemicals
Catalog number456270010
Price (USD)/ Each
428.65
Online exclusive
476.00 
Save 47.35 (10%)
-
Add to cart
Chemical Identifiers
CAS551-68-8
IUPAC Name(3R,4R,5R)-2-(hydroxymethyl)oxane-2,3,4,5-tetrol
Molecular FormulaC6H12O6
InChI KeyLKDRXBCSQODPBY-JDJSBBGDSA-N
SMILESOCC1(O)OC[C@@H](O)[C@@H](O)[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=97.5 %
Water=<2.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only