Thermo Scientific Chemicals

Forchlorfenuron, 98%, Thermo Scientific Chemicals

Catalog number: 456290250
25 g, Each
Thermo Scientific Chemicals

Forchlorfenuron, 98%, Thermo Scientific Chemicals

Catalog number: 456290250
25 g, Each
Quantity
Catalog number: 456290250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
68157-60-8
Appearance (Form)
Powder
Melting point
170°C to 177°C
Infrared spectrum
Conforms
Loss on drying
=<1 %
Appearance (Color)
White to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text