Chemical Identifiers
CAS52315-07-8
IUPAC Namecyano(3-phenoxyphenyl)methyl 3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylate
Molecular FormulaC22H19Cl2NO3
InChI KeyKAATUXNTWXVJKI-UHFFFAOYNA-N
SMILESCC1(C)C(C=C(Cl)Cl)C1C(=O)OC(C#N)C1=CC=CC(OC2=CC=CC=C2)=C1
View more