Thermo Scientific Chemicals

Cypermethrin, 98%, ACROS Organics™

Catalog number: 456310250
25 g, Each
Thermo Scientific Chemicals

Cypermethrin, 98%, ACROS Organics™

Catalog number: 456310250
25 g, Each
Quantity
Catalog number: 456310250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
52315-07-8
IUPAC Name
cyano(3-phenoxyphenyl)methyl 3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylate
Molecular Formula
C22H19Cl2NO3
InChI Key
KAATUXNTWXVJKI-UHFFFAOYNA-N
SMILES
CC1(C)C(C=C(Cl)Cl)C1C(=O)OC(C#N)C1=CC=CC(OC2=CC=CC=C2)=C1
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless to yellow
HPLC
>=95 % (Mixture of isomers)
Infrared spectrum
Conforms
Water
=<1.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text