N-Formyl-L-proline, 97%, Thermo Scientific Chemicals
N-Formyl-L-proline, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Formyl-L-proline, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 456420050
Price (USD)/ Each
457.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
457.00
Add to cart
N-Formyl-L-proline, 97%, Thermo Scientific Chemicals
Catalog number456420050
Price (USD)/ Each
457.00
-
Add to cart
Chemical Identifiers
CAS13200-83-4
IUPAC Name1-formylpyrrolidine-2-carboxylic acid
Molecular FormulaC6H9NO3
InChI KeyDHDRGOURKDLAOT-UHFFFAOYNA-N
SMILESOC(=O)C1CCCN1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder or crystalline powder
Elemental Analysis48.33 to 52.36 % C
Specific optical rotation-115° to -130° (20°C, 589 nm) (c=0.8 in EtOH)
Appearance (Color)White to off-white
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only