Furfuryl isocyanate, 97%, Thermo Scientific Chemicals
Furfuryl isocyanate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Furfuryl isocyanate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 456490010
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
IUPAC Name2-(isocyanatomethyl)furan
Molecular FormulaC6H5NO2
InChI KeyUIADMYLYGJYUSQ-UHFFFAOYSA-N
SMILESO=C=NCC1=CC=CO1
Molecular Weight (g/mol)123.11
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4750 to 1.4770 (20°C, 589 nm)
Appearance (Color)Clear colorless to brown
Appearance (Form)Liquid
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only