Thermo Scientific Chemicals

(R)-3-Methylmorpholine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 456780050
5 g, Each
Thermo Scientific Chemicals

(R)-3-Methylmorpholine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 456780050
5 g, Each
Quantity
Catalog number: 456780050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
953780-78-4
IUPAC Name
3-methylmorpholine hydrochloride
Molecular Formula
C5H12ClNO
InChI Key
MSOCQCWIEBVSLF-UHFFFAOYNA-N
SMILES
Cl.CC1COCCN1
Appearance (Form)
Powder or solid
Appearance (Color)
White to yellow to light brown
Infrared spectrum
Conforms
Titration Argentometric
>=96.0 to =<104.0 %
Specific optical rotation
+1° to +3° (20°C, 589 nm) c=1 in H2O

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text