Thermo Scientific Chemicals

(S)-(+)-p-Toluenesulfinamide, 98%, Thermo Scientific Chemicals

Catalog number: 456800010
1 g, Each
Thermo Scientific Chemicals

(S)-(+)-p-Toluenesulfinamide, 98%, Thermo Scientific Chemicals

Catalog number: 456800010
1 g, Each
Quantity
Catalog number: 456800010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
188447-91-8
IUPAC Name
4-methylbenzene-1-sulfinamide
Molecular Formula
C7H9NOS
InChI Key
YNJDSRPIGAUCEE-UHFFFAOYNA-N
SMILES
CC1=CC=C(C=C1)S(N)=O
Appearance (Color)
White to off-white
NMR (1H-NMR)
Consistent with structure
Infrared spectrum
Conforms
HPLC
>=97.5 %
Optical rotation
+80° to +89° (20°C, 589 nm) (c=1, CHCl3)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text