Thermo Scientific Chemicals

Acetic acid, 25%, Thermo Scientific Chemicals

Catalog number: 456850050
5 L, Each
Thermo Scientific Chemicals

Acetic acid, 25%, Thermo Scientific Chemicals

Catalog number: 456850050
5 L, Each
Quantity
Catalog number: 456850050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
64-19-7
IUPAC Name
acetic acid
Molecular Formula
C2H4O2
InChI Key
QTBSBXVTEAMEQO-UHFFFAOYSA-N
SMILES
CC(O)=O
Appearance (Form)
Solution
Appearance (Color)
Clear colorless
Titration with NaOH
24.0 to 26.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text