Thermo Scientific Chemicals

3,3-Dimethyl-1-(trifluoromethyl)-1,2-benziodoxole, 95%, Thermo Scientific Chemicals

Catalog number: 456900010
1 g, Each
Thermo Scientific Chemicals

3,3-Dimethyl-1-(trifluoromethyl)-1,2-benziodoxole, 95%, Thermo Scientific Chemicals

Catalog number: 456900010
1 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 456900010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
887144-97-0
IUPAC Name
3,3-dimethyl-1-(trifluoromethyl)-3H-1λ³,2-benziodaoxole
Molecular Formula
C10H10F3IO
InChI Key
HVAPLSNCVYXFDQ-UHFFFAOYSA-N
SMILES
CC1(C)O[I](C2=CC=CC=C12)C(F)(F)F
Appearance (Color)
White to light brown
HPLC
>=94 %
Appearance (Form)
Crystalline powder or crystals
Melting point
70°C to 79°C
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text