Thermo Scientific Chemicals

Azomethine H monosodium salt monohydrate, 95%, Thermo Scientific Chemicals

Catalog number: 457090250
25 g, Each
Thermo Scientific Chemicals

Azomethine H monosodium salt monohydrate, 95%, Thermo Scientific Chemicals

Catalog number: 457090250
25 g, Each
Quantity
Catalog number: 457090250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
304655-82-1
IUPAC Name
4-hydroxy-5-{[(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino}naphthalene-2,7-disulfonic acid hydrate sodium
Molecular Formula
C17H15NNaO9S2
InChI Key
CYPHYAYBFNUBEV-UHFFFAOYSA-N
SMILES
O.[Na].OC1=CC(=CC2=CC(=CC(NC=C3C=CC=CC3=O)=C12)S(O)(=O)=O)S(O)(=O)=O
Appearance (Color)
Yellow to orange
Appearance (Form)
Powder
Infrared spectrum
Conforms
Water
=<6 %
Titration with NaOH
94.0 to 106.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text