Thermo Scientific Chemicals

2,4-Dimethyl-1,3-oxazole-5-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: 457350010
1 g, Each
Thermo Scientific Chemicals

2,4-Dimethyl-1,3-oxazole-5-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: 457350010
1 g, Each
Quantity
Catalog number: 457350010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2510-37-4
IUPAC Name
dimethyl-1,3-oxazole-5-carboxylic acid
Molecular Formula
C6H7NO3
InChI Key
JLSFKHJNJFXGAB-UHFFFAOYSA-N
SMILES
CC1=NC(C)=C(O1)C(O)=O
Appearance (Color)
White to off-white
HPLC
>=96 %
Appearance (Form)
Powder
Infrared spectrum
Conforms
Melting point
225°C to 235°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text